missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gilac™ Pedal Container with Lid
Pedal Container with Lid offers hands-free operation for sanitary disposal.
Brand: Gilac™ G614422
This item is not returnable.
View return policy
Description
- Equipped with two antimark wheels (diameter 100 mm)
- Easy to move around
- Hands-free operation for sanitary disposal
- Easy to clean
- Coloured lids for selective waste sorting
Specifications
100 L | |
26.4 gal. | |
515 x 477 x 925 mm |
Blue | |
White | |
Polypropylene |