missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gilac™ Pedal Container with Lid
Pedal Container with Lid offers hands-free operation for sanitary disposal.
£226.00 - £229.00
Specifications
Capacity (Metric) | 100 L |
---|---|
Capacity (English) | 26.4 gal. |
Material | Polypropylene |
Product Code | Brand | Closure Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Closure Color | Price | Quantity & Availability | |||||
11857443
|
Gilac™
G614421 |
White |
£229.00
Each |
Estimated Shipment: 23-12-2024
Log in to see stock. |
|||||
11877443
|
Gilac™
G614423 |
Green |
£226.00
Each |
Estimated Shipment: 23-12-2024
Log in to see stock. |
|||||
11867443
|
Gilac™
G614422 |
Blue |
£226.00
Each |
Estimated Shipment: 23-12-2024
Log in to see stock. |
|||||
11887443
|
Gilac™
G614424 |
£226.00
Each |
Estimated Shipment: 24-02-2025
Log in to see stock. |
||||||
Description
- Equipped with two antimark wheels (diameter 100 mm)
- Easy to move around
- Hands-free operation for sanitary disposal
- Easy to clean
- Coloured lids for selective waste sorting
Specifications
100 L | |
26.4 gal. | |
Polypropylene |