missing translation for 'onlineSavingsMsg'
Learn More

Gilac™ Pedal Container with Lid

Pedal Container with Lid offers hands-free operation for sanitary disposal.

Brand:  Gilac™ G614423

 View more versions of this product

Product Code. 11877443

  • £226.00 / Each
Estimated Shipment: 23-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Equipped with two antimark wheels (diameter 100 mm)
  • Easy to move around
  • Hands-free operation for sanitary disposal
  • Easy to clean
  • Coloured lids for selective waste sorting
Specifications

Specifications

100 L
26.4 gal.
515 x 477 x 925 mm
Green
Green
Polypropylene
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers