missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PEEK Capillary Tubing

Use these sample loops for your Thermo Scientific™ Rheodyne™ HPLC sample injectors.

Brand:  Thermo Scientific™ 37020-20

Code : 16

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10161033

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Use Thermo Scientific™ Rheodyne™ Sample Loops  for your Rheodyne HPLC sample injectors. They are available in stainless steel or biocompatible PEEK.

  • Broad chemical compatibility
  • Biocompatible
  • Easily cut to desired length
  • Appropriate for many HPLC applications
  • Resistant to most organic solvents however nitric acid, sulfuric acid, dichloromethane, THF and DMSO are not recommended
Specifications

Specifications

Rheodyne HPLC Sample Injectors
0.020 in. ID; 20 cm length; Orange
0.020 in.
7.874 in.
PEEK
Capillary Tubing
HPLC
0.508 mm
20 cm
Product Suggestions

Product Suggestions

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.