Learn More
Thermo Scientific™ PEEK Fingertight Fittings
Ensure HPLC quality and robustness with Thermo Scientific™ PEEK Fingertight Fittings.
Brand:  Thermo Scientific™ U-467BLK
1.14 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 15461073
  • £1.95 / Each
Estimated Shipment: 12-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Ensure HPLC quality and robustness with Thermo Scientific™ PEEK Fingertight Fittings. Machined for reliability, these fittings resist cracking, breaking, thread stripping and leaking in both low- and high-pressure applications. Fingertight plumbing ensures ease of use. The fittings are biocompatible for use across a broad range of applications.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Short Head Port Plug, Black
10-32 Coned Port
1 Each
1/16 in., 10-32 Coned, Delrin, Black
Delrin Acetal
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.