Learn More
Cytiva PEEK Stop Plug
Enables connection of HiTrap columns to a syringe, laboratory pump or chromatography system. Cytiva PEEK Stop Plugs are used to seal the top of a chromatography column, e.g. HiTrap™ prepacked columns. Fingertight or standard 1/16 in. or standard 5/16 in. male stop plugs available.
Brand:  Cytiva 11-0003-55
Code : GX
Additional Details : Weight : 0.00500kg
 View more versions of this product
Product Code. 11779555
  • £52.57 / Pack of 5
Estimated Shipment: 05-08-2024
to see stock.
Add to basket
Description
Description
The inlet and outlet design of HiTrap columns eliminates the need for column connectors when used with ÄKTAdesign™ chromatography systems. Connection to a syringe, laboratory pump, or chromatography system such as FPLC™ System is also possible using these connectors.
  • Made of durable polyetheretherketone (PEEK)
  • One fingertight stop plug is connected to each HiTrap column at delivery
  • Available in narrow 1/16 in., standard 1/16 in. and standard 5/16 in. male stop plugs
Specifications
Specifications
Stop Plug
5 Pk.
Chromatography Columns
Narrow Stop Plug 1/16 in. Male
PEEK