Learn More
Cytiva PEEK Stop Plug
Enables connection of HiTrap columns to a syringe, laboratory pump or chromatography system. Cytiva PEEK Stop Plugs are used to seal the top of a chromatography column, e.g. HiTrap™ prepacked columns. Fingertight or standard 1/16 in. or standard 5/16 in. male stop plugs available.
£44.00 - £52.57
Specifications
Type | Stop Plug |
---|---|
Material | PEEK |
For Use With (Equipment) | Chromatography Columns |
Product Code | Brand | Connections | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Connections | Price | Quantity & Availability | |||||
11380052
|
Cytiva
18-1112-52 |
Stop Plug 1/16 in. Male |
£44.00
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
11779555
|
Cytiva
11-0003-55 |
Narrow Stop Plug 1/16 in. Male |
£52.57
Pack of 5 |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||||
Description
The inlet and outlet design of HiTrap columns eliminates the need for column connectors when used with ÄKTAdesign™ chromatography systems. Connection to a syringe, laboratory pump, or chromatography system such as FPLC™ System is also possible using these connectors.- Made of durable polyetheretherketone (PEEK)
- One fingertight stop plug is connected to each HiTrap column at delivery
- Available in narrow 1/16 in., standard 1/16 in. and standard 5/16 in. male stop plugs
Specifications
Stop Plug | |
PEEK | |
Chromatography Columns |