missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan Scientific and Medical PEEK Tubing, 1/32 in. O.D., 10 ft
Designed specifically for modern LC, LCMS and automation useage. A flexible alternative to stainless steel tubing in high pressure applications, it ensures reproducibility in your chromatography flow rates and results through smooth internal surface along with consistent and tight toleranced IDs
Brand: Trajan Scientific and Medical 1301020004-10F
This item is not returnable.
View return policy
Description
- Excellent chemical resistance and inert to most commonly used solvents. Exceptions are very caustic reagents, strong acids and bases
- Bio-inert ad bio-compatible
- Color coded to industry standards for easy identification of inner diameter
- Leach-free colorants
- Smooth inner walls and precise IDs
- Works well with either polymer fittings as well as stainless steel fittings
- Can be formed to specific customer needs
Specifications
Orange | |
0.020 in. | |
10 ft. | |
3000 psi (207 bar) |
2 to 14 | |
1/32 in. | |
PEEK | |
100°C |
For research use only