missing translation for 'onlineSavingsMsg'
Learn More

Trajan Scientific and Medical PEEK Tubing, 1/32 in. O.D., 10 ft

Designed specifically for modern LC, LCMS and automation useage. A flexible alternative to stainless steel tubing in high pressure applications, it ensures reproducibility in your chromatography flow rates and results through smooth internal surface along with consistent and tight toleranced IDs

£59.55 - £87.00

Specifications

Diameter (English) Outer 1/32 in.
Overall Length (English) 10 ft.
Material PEEK
Max. Operating Temperature (Metric) 100°C
View More Specs

Products 8
Product Code Brand Color Diameter (English) Inner Price Quantity & Availability  
Product Code Brand Color Diameter (English) Inner Price Quantity & Availability  
16632552
View Documents
Trajan Scientific and Medical
1301002500-10F
Natural 0.0025 in.
£59.55
Each
Estimated Shipment: 22-01-2025
to see stock.
Add to basket
 
16672552
View Documents
Trajan Scientific and Medical
1301004006-10F
Black 0.004 in.
£61.15
Each
Estimated Shipment: 22-01-2025
to see stock.
Add to basket
 
15780979
View Documents
Trajan Scientific and Medical
1301005001-10F
Red 0.005 in.
£79.40
Each
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
 
15720989
View Documents
Trajan Scientific and Medical
1301007002-10F
Yellow 0.007 in.
£64.20
Each
Estimated Shipment: 22-01-2025
to see stock.
Add to basket
 
15760989
View Documents
Trajan Scientific and Medical
1301010003-10F
Blue 0.010 in.
£79.05
Each
Estimated Shipment: 22-01-2025
to see stock.
Add to basket
 
15700999
View Documents
Trajan Scientific and Medical
1301015008-10F
Gray 0.015 in.
£59.55
Each
Estimated Shipment: 23-01-2025
to see stock.
Add to basket
 
16612562
View Documents
Trajan Scientific and Medical
1301015000-10F
Natural 0.015 in.
£65.00
Each
Estimated Shipment: 22-01-2025
to see stock.
Add to basket
 
15740999
View Documents
Trajan Scientific and Medical
1301020004-10F
Orange 0.020 in.
£87.00
Each
Estimated Shipment: 17-01-2025
to see stock.
Add to basket
 
Description

Description

  • Excellent chemical resistance and inert to most commonly used solvents. Exceptions are very caustic reagents, strong acids and bases
  • Bio-inert ad bio-compatible
  • Color coded to industry standards for easy identification of inner diameter
  • Leach-free colorants
  • Smooth inner walls and precise IDs
  • Works well with either polymer fittings as well as stainless steel fittings
  • Can be formed to specific customer needs
Specifications

Specifications

1/32 in.
10 ft.
PEEK
100°C
Videos
SDS
Documents

Documents

Product Certifications

For research use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.