missing translation for 'onlineSavingsMsg'
Learn More
3M™ PELTOR™ Earmuffs, Yellow Product Code.: 13489918
Add to basket

3M™ PELTOR™ Earmuffs, Yellow

Product Code. 13489918
X2A, Yes, Over-the-head
Click to view available options
Model:
X2A
Dielectric:
Yes
Style:
Over-the-head

Product Code. 13489918

Brand: 3M™ 7000103989

Add to basket
Add to basket

3M™ PELTOR™ Earmuffs are our innovative hearing protectors with a modern slim cup design.

  • Reduces noise levels
  • Colour-coding makes it easy to identify the level of protection being worn
  • Slim cups offer improved compatibility with other safety equipment
  • Wide soft ear cushions help workers stay comfortable
  • Designed to optimise weight distribution and balance for outstanding comfort
  • ABS plastic cups are rigid, high strength and impact resistant
  • Thermoplastic polyurethane (TPU) soft outer cup coating protects from abrasion and bumps

Compliance

3M™ ear muffs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.

TRUSTED_SUSTAINABILITY

Specifications

Color Black, Yellow
ANSI S3.1974 Yes
CSA Z94.2 Class A
Cushion Color Black
For Use With (Application) Assembly, Cleaning, Demolition, Facility Maintenance, Grinding, Machine Operations, Painting, Sanding, Welding
Model X2A
Pairs per Package 1 Ea.
Noise Reduction Rating (NRR) 24 dB
Product Type X2 Series
Headband Color Black
Dielectric Yes
Style Over-the-head
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3M™ PELTOR™ Earmuffs, Yellow >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.