Learn More
UVP Pen-Ray™ Gas Lamp Filll
Brand:  UVP 90-0015-01
 View more versions of this product
Product Code. 12843818
  • £269.00 / Each
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Pen Ray
450 V
117.3 mm
Neon
254 nm
Quartz