missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Pepstatin A

Protease inhibitor, inhibits Pepsin, Renin and Cathepsin D.

£37.75 - £109.11

Chemical Identifiers

CAS 26305-03-3
Molecular Formula C34H63N5O9, C34H63N5O9
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15855328
View Documents Promotion Details
Thermo Scientific Chemicals
J20037.#0
5 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15865328
View Documents Promotion Details
Thermo Scientific Chemicals
J20037.MB
25 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Applications: Pepstatin A, a protease inhibitor, inhibits Pepsin, Renin and Cathepsin D. Effective concentrations are 10-6 to 10-9M. It does not inhibit Trypsin, Chymotrypsin, Papain or Plasmin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Protease inhibitor, inhibits Pepsin, Renin and Cathepsin D
  • Effective concentrations are 10-6 to 10-9M
  • Does not inhibit Trypsin, Chymotrypsin, Papain or Plasmin
  • Protease inhibitor, inhibits Pepsin, Renin and Cathepsin D
  • MDL: MFCD00060740
Specifications

Chemical Identifiers

26305-03-3
C34H63N5O9, C34H63N5O9

Specifications

10-6 to 10-9M
C34H63N5O9, C34H63N5O9
Dissolves in methanol
Powder
CC(C)CC(NC(=O)[C@H](C)NC(=O)CC(O)C(CC(C)C)NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)CC(C)C)C(C)C)C(C)C)C(O)CC(O)=O
685.90
26305-03-3
Isovaleryl-val-val-4-amino-3-hydroxy-6-methylheptanoyl-ala-4-amino-3-hydroxy-6-methylheptanoic acid SDS Certificate of Analysis Product Specification Technical Inquiry
2202362
FAXGPCHRFPCXOO-JHTFMCFMSA-N
3-hydroxy-4-[(2S)-2-{3-hydroxy-6-methyl-4-[(2S)-3-methyl-2-[(2S)-3-methyl-2-(3-methylbutanamido)butanamido]butanamido]heptanamido}propanamido]-6-methylheptanoic acid
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 247-600-0

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only