Learn More
Fisher BioReagents™ Peptone, Granulated
Mixture of peptides and free amino acids from pancreatic hydrolysis of animal tissue. Fisher Bioreagents Peptone, Granulated is an enzymatic digest of meat tissue, nutritious and capable of supporting growth of many fastidious microorganisms.
Brand:  Fisher BioReagents™ 12831660
 View more versions of this product
Product Code. 12831660
  • £502.00 / 2kg
Estimated Shipment: 20-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Soluble in water
  • Light Beige
  • Particle Size: 1-2mm
Specifications
Specifications
2000 g