missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ Peptone, Granulated
Mixture of peptides and free amino acids from pancreatic hydrolysis of animal tissue. Fisher Bioreagents Peptone, Granulated is an enzymatic digest of meat tissue, nutritious and capable of supporting growth of many fastidious microorganisms.
£260.76 - £455.07
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12831660
|
Fisher BioReagents™
12831660 |
2000 g |
£455.07
2kg |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
12841660
|
Fisher BioReagents™
12841660 |
500 g |
£260.76
500g |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
Description
- Soluble in water
- Light Beige
- Particle Size: 1-2mm