Learn More
Fisher BioReagents™ Peptone, Granulated
Mixture of peptides and free amino acids from pancreatic hydrolysis of animal tissue. Fisher Bioreagents Peptone, Granulated is an enzymatic digest of meat tissue, nutritious and capable of supporting growth of many fastidious microorganisms.
Brand: Fisher BioReagents™ 12841660
Code : 32
Additional Details : Weight : 0.60000kg
Description
- Soluble in water
- Light Beige
- Particle Size: 1-2mm
Specifications
500 g |