missing translation for 'onlineSavingsMsg'
Learn More
Perchloric Acid, TraceSELECT™ Ultra, 67 to 72%, Honeywell Fluka™ Product Code.: 15601150

Perchloric Acid, TraceSELECT™ Ultra, 67 to 72%, Honeywell Fluka™

Product Code. 15601150 Shop All Honeywell Fluka Products
250 mL, Poly Bottle
Click to view available options
Quantity:
250 mL
500 mL
1 L
Packaging:
Poly Bottle
This item is not returnable. View return policy

Product Code. 15601150

Brand: Honeywell-Fluka 12415250ML

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

TraceSELECT™ Ultra, for ultratrace analysis, 67-72%

TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions

 


 

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7601-90-3
Molecular Formula ClHO4
Molecular Weight (g/mol) 100.45
MDL Number MFCD00011325
InChI Key VLTRZXGMWDSKGL-UHFFFAOYSA-N
Synonym unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4Show MoreShow Less
PubChem CID 24247
ChEBI CHEBI:29221
IUPAC Name perchloric acid
SMILES O[Cl](=O)(=O)=O

Specifications

Color Colorless
Melting Point -18°C
Boiling Point 203°C
Density 1.66g/cu.cm
Packaging Poly Bottle
Flash Point 113°C (235.4°F)
Quantity 250 mL
Linear Formula HClO4
Physical Form Liquid
UN Number UN1873
Formula Weight 100.46g/mol
Percent Purity 67 to 72%
Grade TraceSELECT™ Ultra
Chemical Name or Material Perchloric Acid
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Perchloric acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Substances and mixtures corrosive to metals Category 1
  • Oxidising liquid Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H271-May cause fire or explosion; strong oxidiser.
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H373-May cause damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P234-Keep only in original container.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Perchloric acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Perchloric Acid, TraceSELECT™ Ultra, 67 to 72%, Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.