missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Percoll™ Media

Well-referenced media for density gradient centrifugation of cells, viruses, and subcellular particles. Cytiva™ Percoll™ Media is composed of colloidal silica coated with polyvinylpyrrolidone (PVP).

Brand:  Cytiva 17-0891-09

Product Code. 11748498

  • £2043.00 / 6 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Composed of colloidal silica coated with polyvinylpyrrolidone (PVP)
  • Used for separation of cells, subcellular particles and larger viruses under gentle conditions which preserve viability and morphological integrity of cells
  • Low viscosity density gradient medium reduces required centrifugal force
  • Nontoxic to cells
  • Compatible to ionic strength and pH
  • Gradients can either be preformed or created by centrifugation at moderate speeds in an angle-head rotor
  • Gradients are iso-osmotic throughout and cover a range of densities up to 1.3g/mL
  • Buffered within the pH range 5.5 to 10.0 without any changes in properties
  • Conductivity (mS/m): max. 100
  • Osmolality (mOsm/kg H2O): max. 25
  • Viscosity (cP): max 15

Order Info

Avaialble in easy-to-open, resealable 250mL and 1L bottles

TRUSTED_SUSTAINABILITY
Specifications

Specifications

6 x 1 L
Cell Isolation
<0.12EU/mL
Colorless to Slightly Yellow
Centrifugation Media
Liquid
4–30°C
15cP
< 25 mOsm/kg
8.5 to 9.5
1.135 g/mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cytiva Percoll™ Media > Density: 1.135g/ml; Shelf life: 5years

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.