missing translation for 'onlineSavingsMsg'
Learn More
Perfluoro(2-methyl-2-pentene), 97%, Thermo Scientific Chemicals Product Code.: 11418327

Perfluoro(2-methyl-2-pentene), 97%, Thermo Scientific Chemicals

Product Code. 11418327
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11418327

Brand: Thermo Scientific Alfa Aesar A18394.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1584-03-8
Molecular Formula C6F12
Molecular Weight (g/mol) 300.047
MDL Number MFCD00015724
InChI Key FAEGGADNHFKDQX-UHFFFAOYSA-N
Synonym perfluoro-2-methyl-2-pentene, perfluoro 2-methylpent-2-ene, hexafluoropropene dimer, perfluoro-2-methylpent-2-ene, 1,1,1,3,4,4,5,5,5-nonafluoro-2-trifluoromethyl pent-2-ene, perfluoro 2-methyl-2-pentene, nonafluoro-2-trifluoromethyl pent-2-ene, 1,1,1,3,4,4,5,5,5-nonafluoro-2-trifluoromethyl pent-4-ene, 2-pentene, 1,1,1,3,4,4,5,5,5-nonafluoro-2-trifluoromethyl, 1,1,1,3,4,4,5,5,5-nonafluoro-2-trifluoromethyl-2-penteneShow MoreShow Less
PubChem CID 74105
IUPAC Name 1,1,1,3,4,4,5,5,5-nonafluoro-2-(trifluoromethyl)pent-2-ene
SMILES C(=C(C(C(F)(F)F)(F)F)F)(C(F)(F)F)C(F)(F)F

Specifications

Density 1.62
Boiling Point 48°C to 54°C
Flash Point −12°C (10°F)
Refractive Index ca. 1.3
Quantity 5 g
UN Number UN1993
Formula Weight 300.05
Percent Purity 97%
Chemical Name or Material Perfluoro(2-methyl-2-pentene)

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10201328 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA18394.14,A18394.06
Lot Number10158623 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18394.14,A18394.06
Lot Number10226374 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18394.14,A18394.06
Lot Number10224093 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18394.14,A18394.06
Lot NumberI9945A Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18394.14,A18394.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Perfluoro(2-methyl-2-pentene), 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.