missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Perfluoro Acid Anhydrides (TFAA, PFAA and HFAA) Acylation Reagents

Prepare volatile and stable fluoracyl derivatives for GC/MS with the Thermo Scientific™ Perfluoro Acid Anhydrides (TFAA, PFAA and HFAA).

Brand:  Thermo Scientific™ TS-63164


 View more versions of this product

Product Code. 11772247

  • £269.00 / Pack of 10

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Prepare fluoracyl derivatives for GC/MS and for flame ionization and electron-capture detection techniques with the Thermo Scientific™ Perfluoro Acid Anhydrides (TFAA, PFAA and HFAA). These reagents react readily with alcohols, phenols, and amines.

  • Used to prepare fluoracyl derivatives for GC/MS
  • Produce stable volatile derivatives for FID and ECD techniques
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GC/MS
10 x 1 mL
98 to 100%
Room Temperature
Heptafluoro-butanoic acid anhydride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Perfluoro Acid Anhydrides

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P363-Wash contaminated clothing before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : heptafluorobutyric

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.