missing translation for 'onlineSavingsMsg'
Learn More

Pericentrin Rabbit anti-Canine, Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 22271-1-AP-20UL

 View more versions of this product

Product Code. 16851815

  • £135.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The antibody is specific to PCNT and it can recognize isoform 1 and 2.

This gene encodes a protein that was identified in a screen for genes expressed in metastatic cells, specifically, mammary adenocarcinoma cell lines. Expression of this gene has been correlated with the metastatic potential of at least two types of carcinomas although it is also expressed in many normal tissues. The role it plays in metastasis is unclear. It was initially thought to be the 70kD component of a nucleosome remodeling deacetylase complex, NuRD, but it is more likely that this component is a different but very similar protein. These two proteins are so closely related, though, that they share the same types of domains. These domains include two DNA binding domains, a dimerization domain, and a domain commonly found in proteins that methylate DNA. The profile and activity of this gene product suggest that it is involved in regulating transcription and that this may be accomplished by chromatin remodeling.
Specifications

Specifications

Pericentrin
Polyclonal
Unconjugated
PCNT
KEN, Kendrin, KIAA0402, MOPD2, PCN, PCNT, PCNT2, PCNTB, PCTN2, pericentrin, Pericentrin B, SCKL4
Rabbit
Antigen Affinity Chromatography
RUO
491613, 5116
-20°C
Liquid
Immunocytochemistry, Immunofluorescence
0.36 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O95613
PCNT
Peptide
20 μL
Primary
Canine, Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers