missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pericentrin Rabbit anti-Canine, Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | Pericentrin |
---|---|
Concentration | 0.36 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16851815
|
Proteintech
22271-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16841815
|
Proteintech
22271-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The antibody is specific to PCNT and it can recognize isoform 1 and 2.
This gene encodes a protein that was identified in a screen for genes expressed in metastatic cells, specifically, mammary adenocarcinoma cell lines. Expression of this gene has been correlated with the metastatic potential of at least two types of carcinomas although it is also expressed in many normal tissues. The role it plays in metastasis is unclear. It was initially thought to be the 70kD component of a nucleosome remodeling deacetylase complex, NuRD, but it is more likely that this component is a different but very similar protein. These two proteins are so closely related, though, that they share the same types of domains. These domains include two DNA binding domains, a dimerization domain, and a domain commonly found in proteins that methylate DNA. The profile and activity of this gene product suggest that it is involved in regulating transcription and that this may be accomplished by chromatin remodeling.Specifications
Pericentrin | |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Canine, Human | |
O95613 | |
491613, 5116 | |
Peptide | |
Primary | |
-20°C | |
PCNT |
0.36 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
KEN, Kendrin, KIAA0402, MOPD2, PCN, PCNT, PCNT2, PCNTB, PCTN2, pericentrin, Pericentrin B, SCKL4 | |
PCNT | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |