missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Perlapine
Potent muscarinc DREADD agonist
£137.15 - £515.11
Specifications
Chemical Name or Material | 6-(4-Methyl-1-piperazinyl)-11 H-dibenz[b,e]azepine |
---|---|
CAS | 1977-11-3 |
Target | DREADD Activators |
Molecular Formula | C19H21N3 |
Formula Weight | 291.39 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18738186
|
Tocris Bioscience™
5549/50 |
50 mg |
£515.11
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18795456
|
Tocris Bioscience™
5549/10 |
10 mg |
£137.15
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potent muscarinic DREADD agonist (pEC50 values are 8.38, 8.08 and 7.27 for hM1Dq, hM3Dq and hM4Di in vitro, respectively). Exhibits >10,000-fold selectivity for hM3Dq over hM3 receptors.Specifications
6-(4-Methyl-1-piperazinyl)-11 H-dibenz[b,e]azepine | |
DREADD Activators | |
291.39 |
1977-11-3 | |
C19H21N3 | |
99% |
Safety and Handling
Recommended Storage : Store at Room Temperature