Learn More
Macherey-Nagel™ Permabond™ SE-52 GC Capillary Column
Brand:  Macherey-Nagel™ 723312.25
 View more versions of this product
Product Code. 10609961
  • £557.00 / Each
Estimated Shipment: 13-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
0.5 mm
0.32 mm
G27, G36
300°C to 320°C
25 m
1 Each
GC Capillary Column
Permabond™ SE-52
Gas Chromatography (GC)
0.5 μm
Permabond™ SE-52