Learn More
Brady™ PermaSleeve™ Bulk Low-smoke, Zero-halogen Heat Shrink Wire and Cable Labels for M7 Printers
PermaSleeve™ Heat Shrink Tubing is engineered for thermal transfer printing and is ideal for use in wire identification.
Brand:  Brady™ 174483
Additional Details : Weight : 0.76300kg
 View more versions of this product
Product Code. 17828854
  • £745.00 / Pack of 1000
Estimated Shipment: 06-09-2024
to see stock.
Add to basket
Description
Description
  • Suitable for use in applications that require low-smoke, halogen-free, flame retardant markers
  • Available in multiple sizes
Specifications
Specifications
ASTM D 2671, ASTM E 162, ASTM E 662, ASTM G 155, MIL-STD-202, SAE-AMS-DTL-23053, SAE-AS-81531, UL Recognized
White
Die-cut
Labels
Data and Telecommunications Labeling, Wire and Cable Labeling
1000 Labels
-67°F to +221°F
8 to 4 AWG
0.645 in.
25.4 mm
0.375 in.
Non-adhesive
Matte
Polyolefin
Non-sterile
M710, BMP71
Rectangle
-55°C to +105°C
0.645 x 1 in.
1 in.
16.38 mm
9.53 mm