Learn More
Brady™ PermaSleeve™ Heat Shrink Wire and Cable Continuous Labels for M6/M7 Printers
Designed for rigorous wire marking applications, PermaSleeve™ Heat Shrink Polyolefin labels combine speed, ease of use, and permanence to create high quality labeling.
Brand:  Brady™ 174507
 View more versions of this product
Product Code. 17858954
  • £889.00 / Each
Estimated Shipment: 11-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Very high temperature resistance, UV resistant, chemical resistant, fuel and oil resistant, self-extinguishing
  • Crisp, clear identification resistant to abrasions and fading
  • 3:1 heat shrink labels fit snugly around wires for maximum insulation protection and permanent identification
  • Made of B-342 polyolefin material
Specifications
Specifications
Method 215, Method 215K, MIL-DTL-23053/5 Class 1, MIL-M-81531, MIL-STD-202, MIL-STD-202F, UL 224, RoHS Compliant to 2011/65/EU (Pass 2015/863), SAE AS23053/5 (Class 1), SAE AS-81531
White
Continuous
Labels
Electrical Labeling, Wire and Cable Labeling
Rectangle
-55°C to +135°C
50 ft. x 1.66 in.
15.24 m
42.16 mm
1 in.
Non-adhesive
Matte
Polyolefin
Non-sterile
M710, BMP71
-67°F to +275°F
2 to 500 MCM
50 ft.
1.66 in.
42.16 mm
25.4 mm