Learn More
Brady™ PermaSleeve™ White Heat Shrink Wire and Cable Die-cut Labels for M6/M7 Printers
Designed for rigorous wire marking applications, PermaSleeve™ Heat Shrink Polyolefin labels combine speed, ease of use, and permanence to create high quality labeling.
Brand:  Brady™ 174480
 View more versions of this product
Product Code. 17848934
  • £818.00 / Pack of 2000
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Very high temperature resistance, UV resistant, chemical resistant, fuel and oil resistant, self-extinguishing
  • Crisp, clear identification resistant to abrasions and fading
  • 3:1 heat shrink labels fit snugly around wires for maximum insulation protection and permanent identification
  • Made of B-342 polyolefin
Specifications
Specifications
ASTM D 2671, ASTM E 162, ASTM E 662, ASTM G 155, MIL-STD-202, SAE-AMS-DTL-23053, SAE-AS-81531, UL 224, RoHS Compliant to 2011/65/EU (Pass 2015/863), SAE AS23053/5 (Class 1)
White
Die-cut
Labels
Electrical Labeling, Wire and Cable Labeling
2000 Labels
-67°F to +275°F
12 to 4 AWG
0.645 in.
22.23 mm
0.375 in.
Non-adhesive
Matte
Polyolefin
Non-sterile
M610, M710, M611, BMP61, BMP71
Rectangle
-55°C to +135°C
0.645 x 0.875 in.
0.875 in.
16.38 mm
9.53 mm