Learn More
Brady™ PermaSleeve™ White Heat Shrink Wire and Cable Die-cut Labels for M6/M7 Printers
Designed for rigorous wire marking applications, PermaSleeve™ Heat Shrink Polyolefin labels combine speed, ease of use, and permanence to create high quality labeling.
£275.00 - £970.00
Specifications
Certifications/Compliance | ASTM D 2671, ASTM E 162, ASTM E 662, ASTM G 155, MIL-STD-202, SAE-AMS-DTL-23053, SAE-AS-81531, UL 224, RoHS Compliant to 2011/65/EU (Pass 2015/863), SAE AS23053/5 (Class 1) |
---|---|
Adhesive Type | Non-adhesive |
Color | White |
Finish | Matte |
Format | Die-cut |
Product Code | Brand | Dimensions (LxW) | Height (Metric) | Wire Gauge | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (LxW) | Height (Metric) | Wire Gauge | Price | Quantity & Availability | |||
17858914
|
Brady™
174498 |
0.182 x 0.5 in. | 4.62 mm | 28 to 20 AWG |
£725.00
Pack of 5000 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||
17838984
|
Brady™
174504 |
0.182 x 0.875 in. | 4.62 mm | 28 to 20 AWG |
£970.00
Pack of 5000 |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||
17859094
|
Brady™
174497 |
0.182 x 1 in. | 4.62 mm | 28 to 20 AWG |
£655.00
Pack of 2500 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||
17828794
|
Brady™
174503 |
0.182 x 1.75 in. | 4.62 mm | 28 to 20 AWG |
£885.00
Pack of 2500 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||
17838234
|
Brady™
174427 |
0.235 x 0.583 in. | 5.97 mm | 22 to 16 AWG |
£407.00
Pack of 3000 |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||
17828234
|
Brady™
174420 |
0.235 x 0.5 in. | 5.97 mm | 22 to 16 AWG |
£303.00
Pack of 2000 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||
17838144
|
Brady™
174426 |
0.235 x 0.875 in. | 5.97 mm | 22 to 16 AWG |
£407.00
Pack of 2000 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||
17888134
|
Brady™
174419 |
0.235 x 1 in. | 5.97 mm | 22 to 16 AWG |
£275.00
Pack of 1000 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||
17868234
|
Brady™
174425 |
0.235 x 1.75 in. | 5.97 mm | 22 to 16 AWG |
£370.00
Pack of 1000 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||
17808314
|
Brady™
174446 |
0.335 x 0.437 in. | 8.51 mm | 20 to 10 AWG |
£453.00
Pack of 4000 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||
17878374
|
Brady™
174438 |
0.335 x 0.515 in. | 8.51 mm | 20 to 10 AWG |
£337.00
Pack of 2000 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||
17898314
|
Brady™
174445 |
0.335 x 0.583 in. | 8.51 mm | 20 to 10 AWG |
£453.00
Pack of 3000 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||
17808264
|
Brady™
174444 |
0.335 x 0.875 in. | 8.51 mm | 20 to 10 AWG |
£453.00
Pack of 2000 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||
17868374
|
Brady™
174437 |
0.335 x 1 in. | 8.51 mm | 20 to 10 AWG |
£307.00
Pack of 1000 |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||
17878384
|
Brady™
174443 |
0.335 x 1.75 in. | 8.51 mm | 20 to 10 AWG |
£412.00
Pack of 1000 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||
17888944
|
Brady™
174460 |
0.439 x 1 in. | 11.15 mm | 16 to 8 AWG |
£372.00
Pack of 1000 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||
17808904
|
Brady™
174465 |
0.439 x 1.75 in. | 11.15 mm | 16 to 8 AWG |
£500.00
Pack of 1000 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||
17858804
|
Brady™
174474 |
0.645 x 0.5 in. | 16.38 mm | 12 to 4 AWG |
£605.00
Pack of 2000 |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||
17848934
|
Brady™
174480 |
0.645 x 0.875 in. | 16.38 mm | 12 to 4 AWG |
£810.00
Pack of 2000 |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||
17819014
|
Brady™
174473 |
0.645 x 1 in. | 16.38 mm | 12 to 4 AWG |
£550.00
Pack of 1000 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||
17828994
|
Brady™
174479 |
0.645 x 1.75 in. | 16.38 mm | 12 to 4 AWG |
£740.00
Pack of 1000 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||
17849014
|
Brady™
174485 |
0.851 x 1 in. | 21.62 mm | 8 to 1 AWG |
£635.00
Pack of 1000 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||
17829024
|
Brady™
174489 |
0.851 x 1.75 in. | 21.62 mm | 8 to 1 AWG |
£855.00
Pack of 1000 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||
17868134
|
Brady™
174410 |
1.66 x 1.75 in. | 42.16 mm | 2 to 500 MCM |
£595.00
Pack of 500 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||
Beschreibung
- Very high temperature resistance, UV resistant, chemical resistant, fuel and oil resistant, self-extinguishing
- Crisp, clear identification resistant to abrasions and fading
- 3:1 heat shrink labels fit snugly around wires for maximum insulation protection and permanent identification
- Made of B-342 polyolefin
Spezifikation
ASTM D 2671, ASTM E 162, ASTM E 662, ASTM G 155, MIL-STD-202, SAE-AMS-DTL-23053, SAE-AS-81531, UL 224, RoHS Compliant to 2011/65/EU (Pass 2015/863), SAE AS23053/5 (Class 1) | |
White | |
Die-cut | |
Labels | |
Electrical Labeling, Wire and Cable Labeling | |
Rectangle | |
-55°C to +135°C |
Non-adhesive | |
Matte | |
Polyolefin | |
Non-sterile | |
M610, M710, M611, BMP61, BMP71 | |
-67°F to +275°F |