Learn More
Brady™ PermaSleeve™ White Low-smoke, Zero-halogen Heat Shrink Wire and Cable Labels for M7 Printers
The B-7641 Heat Shrink Sleeve from Brady™ is a low-smoke, halogen-free identification material used for wire and cable identification.
Brand:  Brady™ 174618
Additional Details : Weight : 0.17500kg
 View more versions of this product
Product Code. 17858834
  • £139.00 / Pack of 100
Estimated Shipment: 06-09-2024
to see stock.
Add to basket
Description
Description
  • Made of halogen-free, flexible flame-retardant, heat-shrinkable polyolefin tubing
  • Offers fire safety properties and minimal smoke emission
  • Compatible with the most stringent requirements in the market today to enhance fire safety needs
  • 2:1 heat-shrink ratio
  • Use with thermal transfer printers and Brady™'s R6000 series halogen-free ribbon
Specifications
Specifications
ASTM D 2671, ASTM E 162, ASTM E 662, ASTM G 155, MIL-STD-202, SAE-AMS-DTL-23053, SAE-AS-81531, UL Recognized
White
Die-cut
Labels
Data and Telecommunications Labeling, Wire and Cable Labeling
100 Labels
-67°F to +221°F
8 to 4 AWG
0.645 in.
44.45 mm
0.375 in.
Non-adhesive
Matte
Polyolefin
Non-sterile
M710, BMP71
Rectangle
-55°C to +105°C
0.645 x 1.75 in.
1.75 in.
16.38 mm
9.53 mm