Learn More
Brady™ PermaSleeve Wire Marker Sleeves for M610, BMP61, M611, M710 and BMP71 Printers
PermaSleeve™ Heat Shrinkable Wire Marker Sleeves for M610, BMP61 and M611 printers. Can also be printed on the BMP™ 71 Label Printer and M710 printer, provided the roll adapter (M71-ADAPT-BK) is used.
Brand:  Brady™ 173981
Additional Details : Weight : 0.07800kg
 View more versions of this product
Product Code. 17871549
  • £75.20 / Pack of 200
Estimated Shipment: 11-09-2024
to see stock.
Description
Description
  • PermaSleeve™ Heat Shrinkable Wire Marker Sleeves for M610, BMP61 and M611 printers.
  • Can also be printed on the BMP™71 Label Printer and M710 printer, provided the roll adapter (M71-ADAPT-BK) is used.
Specifications
Specifications
PermaSleeve Wire Marker Sleeves for M610, BMP61, M611, M710 and BMP71
White
Roll
Wire and Cable Markers
M610, M611, M710 (with media adapter), BMP61, BMP71 (with media adapter)
200 Pk.
-55°C to +135°C
12.70 x 11.28 mm
11.28 mm
12.70 mm
Non-adhesive
Matte
Polyolefin
Non-sterile
2
-67°F to +275°F
16 to 8 AWG
0.444 in.
0.5 in.