missing translation for 'onlineSavingsMsg'
Learn More
Perylene, 99+%, Thermo Scientific Chemicals Product Code.: 10357060

Perylene, 99+%, Thermo Scientific Chemicals

Product Code. 10357060
1 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10357060

Brand: Thermo Scientific Acros 130070010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 198-55-0
Molecular Formula C20H12
Molecular Weight (g/mol) 252.31
MDL Number MFCD00004142
InChI Key CSHWQDPOILHKBI-UHFFFAOYSA-N
Synonym peri-dinaphthalene, perilene, dibenz de,kl anthracene, alpha-perylene, perylen, unii-5qd5427un7, ccris 1231, perylen-1-yl, perylen-2-ylShow MoreShow Less
PubChem CID 9142
ChEBI CHEBI:29861
IUPAC Name perylene
SMILES C1=CC2=C3C(=C1)C4=CC=CC5=C4C(=CC=C5)C3=CC=C2

Specifications

Melting Point 277.0°C to 280.0°C
Color Ochre to Brown
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Beilstein 05, I, 363
Merck Index 15, 7299
Solubility Information Solubility in water: insoluble. Other solubilities: slightly soluble in ether, alcohol,, soluble in benzene, toluene,, very soluble in cs2, chloroform, acetone, very sparingly soluble in petroleum ether
Formula Weight 252.31
Percent Purity 99+%
Physical Form Powder
Chemical Name or Material Perylene
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466367 Certificate TypeCertificate of Analysis Date02/10/2024 Catalog Number130070010,130070050
Lot NumberA0437730 Certificate TypeCertificate of Analysis Date09/11/2023 Catalog Number130070010,130070050
Lot NumberR05J010 Certificate TypeCertificate of Analysis Date05/09/2023 Catalog Number130070010,130070050
Lot NumberA0399995 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number130070010,130070050
Lot NumberA0418396 Certificate TypeCertificate of Analysis Date05/05/2020 Catalog Number130070010,130070050
Product Identifier
  • Perylene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Perylene, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.