missing translation for 'onlineSavingsMsg'
Learn More

IKA™ PETTE Multi 12-Channel Pipette

The PETTE multi is a series of mechanical air-displacement pipettes with 12 channels for precise dispensing in microliter or deep well plates.

Brand:  IKA™ 0020017946

429.87 GBP valid until 2024-12-31
Use promo code "21925" to get your promotional price.



Additional Details : Weight : 0.52000kg

 View more versions of this product

Product Code. 17834394

  • £483.00 / Each
Estimated Shipment: 04-09-2024
to see stock.
Add to basket

Description

Description

  • The IKA PETTE multi 12 Channel Pipette Series has 12 channel pipette options that can hold up to 200 μL of liquid.
  • Color-coding simplifies the rapid selection of the right pipette.
  • The impact-resistant, UV-resistant and chemically resistant conical tip ensures reliable metering in the long-term.
  • Improved ergonomics as a result of replaceable handles in various different shapes and materials.
  • The ergonomic handles ensure the best fit for your hand, allowing for precise and repeatable results.
  • The pipetting volume is easily adjusted by fine or coarse adjustments, and the selected volume can be read off the display in large figures at any time.
  • After the pipetting process, the tip of the pipette is ejected quickly by simply pressing the central multifunction button.
  • The piston and tip cone are made from DLC-coated stainless steel, which is resistent to typical wear.
Specifications

Specifications

ISO 8655
Gray
Ergonomic
12
0.01 μL
Gray
Autoclavable
Reusable
Stainless Steel
10 μL
63 x 119 x 235 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers