missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ PFA Class a Volumetric Flask with Screwcap

Manufactured from PerFluoroAlkoxy (PFA) and calibrated to contain (TC, In). Autoclavable and easy to clean.

Brand:  BRAND™ 36220

107.80 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 10579862

  • £157.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • PFA (PerFluoroAlkoxy) construction
  • PFA screw cap with molded sealing ring
  • Calibrated to contain (TC, In)
  • Autoclavable and easy to clean
  • Error limits according to class A
  • Compliant with DIN EN ISO 104
  • Includes one batch certificate
  • Exposure to temperatures up to 121°C (autoclaving) will not cause permanently exceeded error limits
  • Cleaning up to max. 60 °C is recommended to preserve marks and inscriptions
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DIN EN ISO 104
25 mL
0.04 mL
Autoclavable
PFA
PFA
Volumetric flask
A
Product Suggestions

Product Suggestions

Videos

Videos

  • Brand BLAUBRAND

SDS
Documents

Documents

Brochures

Reference Materials

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BRAND™ PFA Class a Volumetric Flask with Screwcap > Capacity (Metric): 25mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.