Learn More
Masterflex™ PFA Threaded Adapter Barbed Fitting
Providing quick and easy assembly when connecting a NPT male thread to a hose barb.
Brand: Masterflex™ 06383-93
This item is not returnable.
View return policy
Description
- Connect fluid paths without the use of tools
- Slip flexible tubing or hose onto the barb to create a seal that typically holds pressures up to 30 psi (2 bar)
- No leaks, flow restrictions or costly downtime
- Sterilization through autoclaving or ethylene oxide (ETO)
Specifications
Autoclavable | |
1/4 NPTM x 3/8 in. tubing I.D. | |
Autoclave or Ethylene Oxide | |
Barbed Fitting |
PFA | |
1 Each | |
1/4 in. NPTM |
Product Content Correction
Product Title
Masterflex™ PFA Threaded Adapter Barbed Fitting > For 1/4 NPTM x 3/8 in. tubing I.D.
Spot an opportunity for improvement?Share a Content Correction