missing translation for 'onlineSavingsMsg'
Learn More

pH 1.00 Buffer Solution at 25°C, Reagecon
BIOPHARMA

Tested, certified, NIST traceable, and shelf-stable buffer solution.

Brand:  Reagecon 1010525

 View more versions of this product

Product Code. 11434539

  • £25.51 / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • pH Buffer Reference Standard, pH Buffer Reference Material
  • Tested and certified by an ISO17025 Accredited Test Method (INAB Ref: 264T) and NIST traceable.
  • Guaranteed stability throughout its entire shelf life, even after opening the bottle.
Specifications

Specifications

Accuracy ± 0.02 pH units
Liquid
HDPE Bottle GL45
1.00
500 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number101024A1 Certificate TypeCertificate of Analysis Date25/03/2025 Catalog Number1010525
Lot Number101024H1 Certificate TypeCertificate of Analysis Date15/03/2025 Catalog Number1010525
Lot Number101024A1 (1) Certificate TypeCertificate of Analysis Date18/06/2024 Catalog Number1010525
Lot Number101023D1 Certificate TypeCertificate of Analysis Date15/03/2024 Catalog Number1010525
Lot Number101022J1 Certificate TypeCertificate of Analysis Date27/04/2023 Catalog Number1010525
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
pH 1.00 Buffer Solution at 25°C, Reagecon > 500 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.