Learn More
pH 1.00 Buffer Solution at 25°C, Reagecon
Tested, certified, NIST traceable, and shelf-stable buffer solution.
Brand: Reagecon 101025
Code : NEW
Additional Details : CAS Number : 7647-01-0 Weight : 1.00400kg Transport : UN number : 1789 Chem class : 8 Pack group : III
Description
- pH Buffer Reference Standard, pH Buffer Reference Material
- Tested and certified by an ISO17025 Accredited Test Method (INAB Ref: 264T) and NIST traceable.
- Guaranteed stability throughout its entire shelf life, even after opening the bottle.
Specifications
Accuracy ± 0.02 pH units | |
Liquid | |
HDPE Bottle GL45 |
1.00 | |
1 L |