missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mettler Toledo™ pH/Conductivity Meter SevenDirect SD23
The SevenDirect SD23 is an accurate and flexible benchtop meter for the determination of pH and conductivity - either separately or simultaneously. The instrument is the replacement of the SevenCompact and is delivered with an EasyPlace sensor arm.
£1995.00 - £2843.16
Specifications
Accuracy (Conductivity) | 0.5 % |
---|---|
Accuracy (mV) | 0.1 mV |
Accuracy (pH) | 0.002 |
Data Management Details | PC, Printer, USB Stick |
Channels | Dual-channel |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
17501006
|
Mettler Toledo™
30671553 |
SevenDirect SD23, Meter Only |
£1995.00
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||||
17511016
|
Mettler Toledo™
30671567 |
SevenDirect SD23 Kit |
£2667.72
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||||
17521016
|
Mettler Toledo™
30671568 |
SevenDirect SD23 Pure H2O |
£2843.16
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||||
Description
SevenDirect instruments are benchtop meters developed for a wide range of laboratory applications, which provide accurate measurements of pH, ORP and conductivity of almost any sample. The SevenDirect is the successor of the SevenCompact and a great addition to any lab.
- Large color touchscreen provides direct measurement and easy data entry.
- Instructions guide users through the sensor calibration process.
- View detailed measurement information on one measurement screen - or focus on the result only and maximize readability with the uFocus mode.
- Closed connector compartment and replaceable cover keep the instrument safe from dust and spills, even with sensors attached (IP54 in-use).
- The instrument stores data like timestamp, sample and sensor ID with every measurement and prints or exports records automatically.
- A two-level user management protects the most important settings.
- The EasyPlace electrode arm makes placing the sensor in the sample or buffer container easy.
- Dedicated positions for cleaning and storage support correct sensor handling.
Specifications
0.5 % | |
0.002 | |
Dual-channel | |
IP54 | |
-2000 to 2000 mV | |
-30°C to +130°C | |
1, 0.1 mV | |
±0.1°C between 0°C and 100°C | |
0.1°C | |
1.9 lb. |
0.1 mV | |
PC, Printer, USB Stick | |
Color TFT | |
0.001 μS/cm to 2000 mS/cm | |
0.1, 0.01, 0.001 | |
2000 Standard Measurements | |
1, 0.1 | |
-22°F to +266°F | |
195 x 205 x 65 mm | |
850 g |