Learn More
Macherey Nagel pH-Fix pH Indicator Strips
Highly appreciated standard strips for many pH testing applications.
Brand: Macherey Nagel 92190
Code : NEW
Additional Details : Weight : 0.30000kg
Description
Highly appreciated standard strips for many pH testing applications.
- Easy dip and read procedure provides reliable results in seconds
- Due to the patented pH-Fix technology, the indicator is chemically bound to the test pad
- Dye does not wash out, so clothes stay clean and samples remain pure
Specifications
1.7 to 3.8 | |
pH Indicator Paper | |
6 mm |
100 Strips | |
85 mm |