Learn More
Macherey-Nagel™ pH-Fix pH Indicator Strips
Highly appreciated standard strips for many pH testing applications.
Brand:  Macherey-Nagel™ 92118
 View more versions of this product
Product Code. 15364184
  • £59.30 / 100 units
Explore more special offers
Description
Description
Highly appreciated standard strips for many pH testing applications.
  • Easy dip and read procedure provides reliable results in seconds
  • Due to the patented pH-Fix technology, the indicator is chemically bound to the test pad
  • Dye does not wash out, so clothes stay clean and samples remain pure
Specifications
Specifications
2 to 9
100 Strips
85 mm
Strip
pH Indicator Paper
6 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Macherey-Nagel™ pH-Fix pH Indicator Strips > pH 2.0 to 9.0 CE marked