Learn More
Fisherbrand™ pH-Fix Test Strips, 1.7-3.8
Provides outstanding measurements safely over the designated pH range. Fisherbrand™ pH-Fix Test Strips, 1.7-3.8 are sufficiently long to avoid contact between the fingers and the test sample.
Brand:  Fisherbrand™ 92190.204231
5.61 GBP valid until 2025-04-18
Use promo code "24095" to get your promotional price.
Product Code. 10017950
  • £40.80 / Pack of 100
Edge
Explore more special offers
Description
Description
  • Available in a flat box so they fit into your pocket
  • Color scale on the test strip box allows exact comparison with the color developing on the test strip pad
  • Hydrophobic plastic handle protects user from sample and allows pH testing in narrow and deep vessels
  • Indicator is covalently bonded on the pads and does not bleed into the sample - can be left in the sample for an indefinite period of time without contamination
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Special Offers
Special Offers
  • Multiple Manufacturers
    Use code 24095 to receive this promotion.
    Promo Code: 24095
    Expires2025-04-15
    Days
    8
    Hours
    11
    Minutes
    29
    Seconds
    6
    Hot Deal
    *Discount will apply to qualifying orders received by Fisher Scientific between 17/03/2025 and 15/04/2025 subject to availability of the product in stock. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy or if the product is no longer available in stock. Other restrictions may apply.
    Terms & Conditions Learn More
  • Multiple Manufacturers
    Browse exclusive prices for our academic customers
    Promo Code: 25081
    Expires2025-12-31
Product Content Correction
Product Title
Fisherbrand™ pH-Fix Test Strips, 1.7-3.8 > pH-Fix Test Strips