missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ pH/Ion Meter SevenDirect SD50
GREENER_CHOICE

Product Code. 17550996 Shop All Mettler Toledo Products
Click to view available options
Probe Type:
Expert Pro-ISM
Micro Pro-ISM
Not included
Pure Pro-ISM
Routine Pro-ISM
Science Pro-ISM
perfectION Fluoride
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17550996

Brand: Mettler Toledo™ 30671548

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

The SevenDirect SD50 is an accurate and flexible benchtop meter for the determination of pH, ORP and ion concentrations. The instrument is the replacement of the SevenCompact and is delivered with an EasyPlace sensor arm.

SevenDirect instruments are benchtop meters developed for a wide range of laboratory applications, which provide accurate measurements of pH, ORP and ion content of almost any sample. The SevenDirect is the successor of the SevenCompact and a great addition to any lab.

  • Large color touchscreen provides direct measurement and easy data entry.
  • Instructions guide users through the sensor calibration process.
  • View detailed measurement information on one measurement screen - or focus on the result only and maximize readability with the uFocus mode.
  • Closed connector compartment and replaceable cover keep the instrument safe from dust and spills, even with sensors attached (IP54 in-use).
  • The instrument stores data like timestamp, sample and sensor ID with every measurement and prints or exports records automatically.
  • A two-level user management protects the most important settings.
  • The EasyPlace electrode arm makes placing the sensor in the sample or buffer container easy.
  • Dedicated positions for cleaning and storage support correct sensor handling.
TRUSTED_SUSTAINABILITY

Specifications

Accuracy (Ion) 0.5 %
Accuracy (mV) 0.1 mV
Accuracy (pH) 0.002
Data Management Details PC, Printer, USB Stick
Description SevenDirect SD50 Pure H2O
Channels 1
Display Type Color TFT
IP Rating IP54
Measurement Modes pH
Range (mV) -2000 to 2000 mV
Range (pH) -2.000 to 20.000
Resolution (pH) 0.1, 0.01, 0.001
Temperature Range (Metric) -30°C to +130°C
Includes InLab Pure-Pro ISM pH/Temperature Sensor
Memory 2000 Standard Measurements
Probe Type Pure Pro-ISM
Product Type Ready-to-measure kit
Range (Ion) -2,000 to 20,000 pX, 0 to 999,999 mg/L (ppm)
Resolution (Ion) 3 or 4 digits (auto-range)
Resolution (mV) 1, 0.1 mV
Resolution (ORP) 1, 0.1
Temperature Accuracy ±0.1°C between 0°C and 100°C
Temperature Range (English) -22°F to +266°F
Temperature Resolution 0.1°C
Dimensions (L x W x H) 195 x 205 x 65 mm
Weight (English) 1.9 lb.
Weight (Metric) 850 g
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.