Learn More
pH Singles Calibration Solutions in 20mL Pouches, Oakton™
Economical pouches are accurate and convenient.
£76.71 - £89.30
Specifications
Quantity 20 mL Pouches
View More Specs
Products 2
Product Code Brand pH Price Quantity & Availability  
Product Code Brand pH Price Quantity & Availability  
12951678
View Documents
Thermo Scientific Eutech
35653-01_CLEAN
4.01
£89.30
Pack of 20
11965484
View Documents Promotion Details
Thermo Scientific Eutech
01X223103
10.00
£76.71
20mL
Description
Description
The pH Singles are ideal for field calibration—just select the pouches needed and carry them to your test site for on-the-spot calibration and verification. No bottles to carry or clean. Pouches are air-tight for maximum freshness and unquestionable reliability. Packets are labeled with temperature versus pH reference tables. Sold in packs of twenty 20mL pouches.
Certifications
NIST traceable, All solutions are certified traceable to NIST™ standards.
Specifications
Specifications
20 mL Pouches