missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Christeyns™ Phago'Derm ASEPT Hand Soap
Phago'Derm ASEPT is a disinfectant cleansing foam soap, specially formulated for hygienic hand washing before any aseptic procedure (insertion of catheters, etc.) and systematically before and after any contact with a patient or contaminating material.
£157.00 - £187.00
Specifications
Product Type | Anti-bacterial Foaming Hand Soap |
---|---|
Color | Light Beige |
Form | Foam |
Product Code | Brand | Volume (Metric) | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Quantity | Price | Quantity & Availability | ||||||
17870245
|
Christeyns™
C65120 |
500 mL | 20 Square Mounted Pump Bottles | N/A | N/A | N/A | |||||
17830265
|
Christeyns™
C65122 |
1000 mL | 12 Airless Bottles |
£187.00
12 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
17880245
|
Christeyns™
C65121 |
1000 mL | 25 Mounted Pump Bottles |
£157.00
12 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
- Formula keeps epidermis soft, even in the event of frequent washing
- Enriched with glycerin
- Specially formulated for the medical, food and institutional sectors
Specifications
Anti-bacterial Foaming Hand Soap | |
Light Beige | |
Foam |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- PHAGO'DERM ASEPT
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : isopropanol, 1-Propanaminium
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Christeyns™ Phago'Derm ASEPT Hand Soap