missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Christeyns™ Phago'Spore Disinfectant Detergent
Phago'Spore is a disinfectant detergent product for all surfaces, including the surfaces of non-invasive medical devices. Phago'Spore is formulated from an active substance (peracetic acid), guaranteeing broad-spectrum microbiological efficacy while ensuring respect for the environment through its biodegradability into acetic acid, oxygen and water.
£49.35 - £94.60
Specifications
Product Type | Sporicide/Disinfectant |
---|---|
Color-coded | No |
Form | Clear Liquid |
pH | 2.1 to 2.3 |
Scent | Fresh Scent |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17860255
|
Christeyns™
C65003 |
750 mL |
£49.35
4.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17870265
|
Christeyns™
C65007 |
5 L |
£94.60
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Ready to use
- Bactericidal, fungicidal, virucidal and sporicidal (active in 10 min. on C. difficile and in 5 min. on Norovirus)
- TP02, TP04 biocide
- Eco-friendly (active substance: peracetic acid)
- 'Food contact' product (in accordance with the decree of September 8, 1999) if used with rinsing
- Dense foam spray head to prevent aerosol formation and protect the user
Specifications
Sporicide/Disinfectant | |
Clear Liquid | |
Fresh Scent |
No | |
2.1 to 2.3 | |
Non-sterile |
Safety and Handling
Product Identifier
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- PHAGO'SPORE
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : waterstofperoxide inoplossing, azijnzuur, Propan-2-ol, Fosforzuur, perazijnzuur
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Christeyns™ Phago'Spore Disinfectant Detergent