missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenethyltrichlorosilane, 95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Specifications
Specifications
CAS | 940-41-0 |
Color | Colorless to Yellow |
Density | 1.2400g/mL |
Boiling Point | 93°C to 96°C (3.0 mmHg) |
Flash Point | 91°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 94% min. (GC) |
Packaging | Glass bottle |
Molecular Formula | C8H9Cl3Si |
Refractive Index | 1.5175 to 1.5195 (20°C, 589nm) |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0394754 | Certificate of Analysis | 08/03/2024 |
Safety and Handling
Product Identifier
- Phenethyltrichlorosilane
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- EUH014-Reacts violently with water.
- EUH029-Contact with water liberates toxic gas.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Phenethyltrichlorosilane, 95%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction