missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenol, 99+%, for biochemistry, loose crystals, Thermo Scientific Chemicals
15.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 39.0°C to 42.0°C |
Color | Colorless to White |
Density | 1.0700g/mL |
Boiling Point | 182.0°C |
Flash Point | 79°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% min. (GC) |
Packaging | Glass bottle |
Linear Formula | C6H5OH |
Quantity | 100 g |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS05_Corrosive.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS06_Toxic.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS08_Dangerous_Health_Hazard.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS09_Aquatic_Toxicity.png)
Product Identifier
- Phenol
- Danger
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- Germ cell mutagenicity Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity after repeated exposure Category 2
- H314-Causes severe skin burns and eye damage.
- H341-Suspected of causing genetic defects.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H301-Toxic if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'