man

Test

CAS RN 108-95-2

OH

CAS RN 108-95-2

IUPAC Name: phenol
Synonyms: phenol carbolicum acidum acide carbolique phenol product caswell no. 649 phenole phenol molten baker P and S liquid and ointment phenol for disinfection phenolated water for disinfection
Molecular Weight (g/mol): 94.11
Molecular Formula: C6H6O
InChi Key: ISWSIDIOOBJBQZ-UHFFFAOYSA-N
SMILES: OC1=CC=CC=C1
Quantity 
  • (4)
  • (2)
  • (2)
  • (1)
  • (7)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (10)
  • (3)
Molecular Weight (g/mol) 
  • (46)
  • (1)
Percent Purity 
  • (5)
  • (8)
  • (6)
  • (3)
  • (2)
  • (4)
Physical Form 
  • (6)
  • (1)
  • (5)
  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
Boiling Point 
  • (8)
  • (7)
  • (9)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (9)
  • (2)
  • (6)
  • (2)
  • (1)
  • (1)
  • (12)
  • (8)
  • (1)
  • (1)
  • (4)
  • (2)
  • (2)
  • (1)
  • (7)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (10)
  • (3)
  • (46)
  • (1)
  • (5)
  • (8)
  • (6)
  • (3)
  • (2)
  • (4)
  • (6)
  • (1)
  • (5)
  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (8)
  • (7)
  • (9)
  • (2)
  • (1)
  • (3)
  • (7)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)