Learn More
Phenol, 99%, extra pure, Thermo Scientific Chemicals
Shop All Thermo Scientific ProductsDescription
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 39.0°C to 42.0°C |
Color | Colorless |
Density | 1.0700g/mL |
Boiling Point | 182.0°C |
Flash Point | 79°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (GC) |
Packaging | Glass bottle |
Linear Formula | C6H5OH |
Quantity | 50 g |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0461999 | Certificate of Analysis | 18/04/2024 | 149340010,149340051,149340500,149340025 |
A0461349 | Certificate of Analysis | 20/03/2024 | 149340010,149340051,149340500,149340025 |
A0459713 | Certificate of Analysis | 29/01/2024 | 149340010,149340051,149340500,149340025 |
A0458509 | Certificate of Analysis | 08/12/2023 | 149340010,149340051,149340500,149340025 |
A0450802 | Certificate of Analysis | 02/05/2023 | 149340010,149340051,149340500,149340025 |
Safety and Handling




- Phenol
- Danger
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- Germ cell mutagenicity Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity after repeated exposure Category 2
- H314-Causes severe skin burns and eye damage.
- H341-Suspected of causing genetic defects.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H301-Toxic if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Special Offers
-
Multiple Manufacturers
Chemical Extravaganza
Shop now and save up to 97% on top-quality products for your lab needs.Promo Code: 25908Expires: 2025-03-31
Days
1Hours
12Minutes
59Seconds
1
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.