missing translation for 'onlineSavingsMsg'
Learn More

Phenol, Fused Crystal, White, Ultrapure Bioreagent, J.T. Baker™

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

£151.00 - £421.00

Chemical Identifiers

CAS 108-95-2
Molecular Formula C6H6O
Molecular Weight (g/mol) 94.11
MDL Number MFCD00002143
InChI Key ISWSIDIOOBJBQZ-UHFFFAOYSA-N
Synonym carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol
PubChem CID 996
ChEBI CHEBI:15882
IUPAC Name phenol
SMILES OC1=CC=CC=C1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15578224
View Documents
Avantor J.T.Baker
4056-00
100 g Steel Can This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15588224
View Documents
Avantor J.T.Baker
4056-01
500 g Steel Can
£421.00
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

    For Extraction of Nucleic Acids and Molecular Biology Applications
Specifications

Chemical Identifiers

108-95-2
94.11
ISWSIDIOOBJBQZ-UHFFFAOYSA-N
996
phenol
C6H6O
MFCD00002143
carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol
CHEBI:15882
OC1=CC=CC=C1

Specifications

108-95-2
79.00°C
MFCD00002143
carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol
OC1=CC=CC=C1
94.11
CHEBI:15882
Ultrapure Bioreagent
Phenol
White
C6H6O
1L = 1.06kg
ISWSIDIOOBJBQZ-UHFFFAOYSA-N
phenol
996
94.11
Fused Crystals
Videos
SDS
Documents

Documents

Product Certifications