Learn More
Corning™ Phenolic Screw Threaded Caps
These caps are fabricated of a resin specially formulated to resist the effects of temperature and steam in autoclaving. Caps have a glued-in, white rubber liner.
Brand:  Corning™ 9999-132
 View more versions of this product
Product Code. 10427033
  • £91.28 / Pack of 288
Estimated Shipment: 02-09-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
These caps are fabricated of a resin specially formulated to resist the effects of temperature and steam in autoclaving. Caps have a glued-in, white rubber liner.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
13-415
Screw Cap
Phenolic
13.5mm
36 Pack
36 Pack
Rubber
Autoclavable
Black
36 Pack
36 Pack
Culture Tubes
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Corning™ Phenolic Screw Threaded Caps > Height: 13.5mm