missing translation for 'onlineSavingsMsg'
Learn More
Phenyl acrylate, 97%, Thermo Scientific Chemicals Product Code.: 11372637

Phenyl acrylate, 97%, Thermo Scientific Chemicals

Product Code. 11372637
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11372637

Brand: Thermo Scientific Alfa Aesar L11819.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Phenyl acrylate reacts with benzaldehyde to produce 2-(hydroxy-phenyl-methyl)-acrylic acid phenyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyl acrylate reacts with benzaldehyde to produce 2-(hydroxy-phenyl-methyl)-acrylic acid phenyl ester.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents. Store at 4°C. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 937-41-7
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.161
MDL Number MFCD00048145
InChI Key WRAQQYDMVSCOTE-UHFFFAOYSA-N
Synonym phenyl acrylate, phenylacrylate, 2-propenoic acid, phenyl ester, phenol acrylate, phenyl-acrylate, acrylic acid phenyl ester, phenyl acrylate 5g, 2-propenoic acid,phenyl ester, acrylic acid phenyl ester, stabilized with bhtShow MoreShow Less
PubChem CID 61242
IUPAC Name phenyl prop-2-enoate
SMILES C=CC(=O)OC1=CC=CC=C1

Specifications

Density 1.076
Boiling Point 89°C to 90°C (12 mmHg)
Refractive Index 1.521
Quantity 5 g
UN Number UN3082
Beilstein 2041124
Solubility Information Insoluble in water.
Formula Weight 148.16
Percent Purity 97%
Chemical Name or Material Phenyl acrylate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberE26W029 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL11819.14,L11819.06
Lot NumberS03B030 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL11819.14,L11819.06
Lot NumberP27E121 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL11819.14,L11819.06
Lot NumberI11U037 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL11819.14,L11819.06
Lot NumberF09R030 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL11819.14,L11819.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Phenyl acrylate, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.